Product Details

SNP ID
rs11147252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133141921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACTCAAATCAATAAAAGAGAAA[A/C]TCTCAAAAGCAGCCAGAAGGAAAAT
Phenotype
MIM: 194538
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF10 PubMed Links

Gene Details

Gene
ZNF10
Gene Name
zinc finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015394.4 Intron NP_056209.2

View Full Product Details