Product Details

SNP ID
rs12139795
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:207457860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTTTCTTCATTTCTTCTTCCT[A/G]AAAAACCTCAACTCATCTTGTCTCA
Phenotype
MIM: 120650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CR2 PubMed Links

Gene Details

Gene
CR2
Gene Name
complement component 3d receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006658.2 Intron NP_001006659.1
NM_001877.4 Intron NP_001868.2
XM_011509206.2 Intron XP_011507508.1

View Full Product Details