Product Details

SNP ID
rs13343673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54537144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTATAAGCCCCCAGTGACCCG[C/T]TGCAACTTTACACCACAGGTGAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIR3DX1 PubMed Links

Gene Details

Gene
KIR3DX1
Gene Name
killer cell immunoglobulin like receptor, three Ig domains X1
There are no transcripts associated with this gene.

View Full Product Details