Product Details

SNP ID
rs12389196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44537155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAAAAACTTCAAAGACTACGTG[G/T]ACTAAATGCGGAAAAGTGGGAGGTG
Phenotype
MIM: 300871
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FUNDC1 PubMed Links

Gene Details

Gene
FUNDC1
Gene Name
FUN14 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173794.3 Intron NP_776155.1

View Full Product Details