Product Details

SNP ID
rs13189159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65669390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGTATATTTATATTTGATAGTTT[A/C]CAAGGTAAGGTTACCATTGGGGAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SGTB PubMed Links

Gene Details

Gene
SGTB
Gene Name
small glutamine rich tetratricopeptide repeat containing beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019072.2 2127 UTR 3 NP_061945.1
XM_005248548.3 2127 UTR 3 XP_005248605.1
XM_017009597.1 2127 UTR 3 XP_016865086.1
Gene
TRAPPC13
Gene Name
trafficking protein particle complex 13
There are no transcripts associated with this gene.

View Full Product Details