Product Details

SNP ID
hCV30997880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:153678747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACACACTCCTCTTTCCTCCCTC[G/T]CGCGCCCTCTCTCATCCTTCTTCAC
Phenotype
MIM: 603181 MIM: 108960
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
There are no transcripts associated with this gene.

Gene
NPR1
Gene Name
natriuretic peptide receptor 1
There are no transcripts associated with this gene.

View Full Product Details