Product Details

SNP ID
rs7741549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10769363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAAGACCATTCATGGTCAAATCT[G/T]ATTGGCTTAAGTACAGAATCATCAC
Phenotype
MIM: 154235
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAK PubMed Links

Gene Details

Gene
MAK
Gene Name
male germ cell associated kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242385.1 Intron NP_001229314.1
NM_001242957.2 Intron NP_001229886.1
NM_005906.5 Intron NP_005897.1
XM_011514619.2 Intron XP_011512921.1
XM_011514620.2 Intron XP_011512922.1
XM_011514621.2 Intron XP_011512923.1
XM_011514622.2 Intron XP_011512924.1
XM_011514624.2 Intron XP_011512926.1
XM_017010863.1 Intron XP_016866352.1
XM_017010864.1 Intron XP_016866353.1
XM_017010865.1 Intron XP_016866354.1
XM_017010866.1 Intron XP_016866355.1
Gene
TMEM14B
Gene Name
transmembrane protein 14B
There are no transcripts associated with this gene.

View Full Product Details