Product Details

SNP ID
rs12401909
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:2233479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGAATTCCCAGGGGTCTTGCA[G/T]GTCCCCTTGCCCTTGCGCTGTGTGG
Phenotype
MIM: 164780
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SKI PubMed Links

Gene Details

Gene
SKI
Gene Name
SKI proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003036.3 Intron NP_003027.1
XM_005244775.3 Intron XP_005244832.1
XM_005244776.4 Intron XP_005244833.1
XM_017002128.1 Intron XP_016857617.1

View Full Product Details