Product Details
- SNP ID
-
rs12337780
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:105562649 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAAGGCAGTTTAGTTTTGGAAAAG[A/G]TTACTATCATCCTTGCTTTAGGTTA
- Phenotype
-
MIM: 607440
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FKTN
PubMed Links
Gene Details
- Gene
- FKTN
- Gene Name
- fukutin
View Full Product Details