Product Details
- SNP ID
-
rs8138123
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36967923 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAAGGCAAGCTTTTCTGGGAAGGG[A/G]CATGAAGACCTTGACATAGTCTGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LL22NC01-81G9.3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8138033] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LL22NC01-81G9.3
- Gene Name
- uncharacterized protein FLJ39582-like
There are no transcripts associated with this gene.
View Full Product Details