Product Details

SNP ID
rs12255832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121958762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTCCCTATTTTTTATTAAAAAC[A/G]AAAACAAAAACAAAACCCACCAAAC
Phenotype
MIM: 612987
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATE1-AS1 PubMed Links

Gene Details

Gene
ATE1-AS1
Gene Name
ATE1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NSMCE4A
Gene Name
NSE4 homolog A, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167865.1 Intron NP_001161337.1
NM_017615.2 Intron NP_060085.2
XM_005269929.4 Intron XP_005269986.1
XM_005269930.3 Intron XP_005269987.1
XM_011539910.2 Intron XP_011538212.1
XM_017016359.1 Intron XP_016871848.1

View Full Product Details