Product Details

SNP ID
rs7625579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69100874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTAGAGAGCAGTGTGGACTGG[A/G]GCTCAATGGAGACTGTTTATTCTGT
Phenotype
MIM: 605709 MIM: 616112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP5 PubMed Links

Gene Details

Gene
ARL6IP5
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006407.3 Intron NP_006398.1
Gene
LMOD3
Gene Name
leiomodin 3
There are no transcripts associated with this gene.

View Full Product Details