Product Details

SNP ID
rs11038852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46292340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCTACAGAAAAAAGGGCCTGA[C/G]TCCATAAGCTCTCTGACACACTCCT
Phenotype
MIM: 616215
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CREB3L1 PubMed Links

Gene Details

Gene
CREB3L1
Gene Name
cAMP responsive element binding protein 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052854.3 Intron NP_443086.1
XM_006718380.3 Intron XP_006718443.1

View Full Product Details