Product Details

SNP ID
rs12049899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69661548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCCTCTGTCCTCACAGGGCTA[A/T]CGGCCCCATGTCTTCACGTGGCAGT
Phenotype
MIM: 168461 MIM: 607224
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCND1 PubMed Links

Gene Details

Gene
CCND1
Gene Name
cyclin D1
There are no transcripts associated with this gene.

Gene
ORAOV1
Gene Name
oral cancer overexpressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153451.2 5431 Intron NP_703152.1
XM_006718470.2 5431 UTR 3 XP_006718533.1

View Full Product Details