Product Details

SNP ID
rs13376280
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:243220863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGAAATAAGAAAAAATTCACTT[C/T]AGTAACACAAAAATATATTCAAAGT
Phenotype
MIM: 613023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP170 PubMed Links

Gene Details

Gene
CEP170
Gene Name
centrosomal protein 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042404.1 Intron NP_001035863.1
NM_001042405.1 Intron NP_001035864.1
NM_014812.2 Intron NP_055627.2
XM_006711843.3 Intron XP_006711906.1
XM_011544334.2 Intron XP_011542636.1
XM_011544335.2 Intron XP_011542637.1
XM_011544336.2 Intron XP_011542638.1
XM_011544337.2 Intron XP_011542639.1
XM_011544338.2 Intron XP_011542640.1
XM_011544339.2 Intron XP_011542641.1
XM_011544340.2 Intron XP_011542642.1
XM_011544341.2 Intron XP_011542643.1
XM_011544342.2 Intron XP_011542644.1
XM_011544343.2 Intron XP_011542645.1
XM_011544344.2 Intron XP_011542646.1
XM_017002932.1 Intron XP_016858421.1
XM_017002933.1 Intron XP_016858422.1
XM_017002934.1 Intron XP_016858423.1
XM_017002935.1 Intron XP_016858424.1
XM_017002936.1 Intron XP_016858425.1
XM_017002937.1 Intron XP_016858426.1
XM_017002938.1 Intron XP_016858427.1
XM_017002939.1 Intron XP_016858428.1
XM_017002940.1 Intron XP_016858429.1
XM_017002941.1 Intron XP_016858430.1
XM_017002942.1 Intron XP_016858431.1
XM_017002943.1 Intron XP_016858432.1
XM_017002944.1 Intron XP_016858433.1
XM_017002945.1 Intron XP_016858434.1
XM_017002946.1 Intron XP_016858435.1
XM_017002947.1 Intron XP_016858436.1
XM_017002948.1 Intron XP_016858437.1
XM_017002949.1 Intron XP_016858438.1
XM_017002950.1 Intron XP_016858439.1
XM_017002951.1 Intron XP_016858440.1

View Full Product Details