Product Details

SNP ID
rs13227435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:114923101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGCGCGTGGCCGAGGCGGGCG[C/G]CGGCCAGCTGGGCTCCACAGCCCAG
Phenotype
MIM: 614511
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MDFIC PubMed Links

Gene Details

Gene
MDFIC
Gene Name
MyoD family inhibitor domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166345.1 658 Missense Mutation GCC,GGC A23G NP_001159817.1
NM_001166346.1 658 Missense Mutation GCC,GGC A132G NP_001159818.3
NM_199072.4 658 Missense Mutation GCC,GGC A132G NP_951038.1

View Full Product Details