Product Details

SNP ID
rs7307228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8535479 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGACTTAATACCCGGGTCATGT[G/T]ATGATTTCTGCAGCAAACCAAACCA
Phenotype
MIM: 609962
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC4E PubMed Links

Gene Details

Gene
CLEC4E
Gene Name
C-type lectin domain family 4 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014358.3 Intron NP_055173.1
XM_011520614.2 Intron XP_011518916.1
XM_011520615.1 Intron XP_011518917.1

View Full Product Details