Product Details

SNP ID
rs12939677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79934446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGGCCCCTGAGCCACCAGGTT[C/T]CTCCATCTTCATGTGGCCACCACGG
Phenotype
MIM: 616637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928766 PubMed Links

Gene Details

Gene
LOC101928766
Gene Name
uncharacterized LOC101928766
There are no transcripts associated with this gene.

Gene
TBC1D16
Gene Name
TBC1 domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271844.1 8768 UTR 3 NP_001258773.1
NM_001271845.1 8768 UTR 3 NP_001258774.1
NM_001271846.1 8768 Intron NP_001258775.1
NM_019020.3 8768 UTR 3 NP_061893.2
XM_005257049.2 8768 UTR 3 XP_005257106.1
XM_005257050.3 8768 UTR 3 XP_005257107.1
XM_005257052.3 8768 Intron XP_005257109.1
XM_006721694.3 8768 UTR 3 XP_006721757.1
XM_006721695.3 8768 Intron XP_006721758.1
XM_011524322.2 8768 UTR 3 XP_011522624.1
XM_011524323.2 8768 UTR 3 XP_011522625.2
XM_011524324.2 8768 UTR 3 XP_011522626.1
XM_017024185.1 8768 Intron XP_016879674.1
XM_017024186.1 8768 UTR 3 XP_016879675.1
XM_017024187.1 8768 UTR 3 XP_016879676.1
XM_017024188.1 8768 UTR 3 XP_016879677.1

View Full Product Details