Product Details

SNP ID
rs10921202
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:192577922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAAAGATGGTTTTATATTCAGT[G/T]GAGCAGAATTTCAATTCAAACCGCG
Phenotype
MIM: 600323
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RGS1 PubMed Links

Gene Details

Gene
RGS1
Gene Name
regulator of G-protein signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002922.3 Intron NP_002913.3

View Full Product Details