Product Details

SNP ID
rs8191053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82035442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAGAATGAGCACTTTCTTCTC[A/G]GACACAGCATGGATCTGCCTGGCTG
Phenotype
MIM: 109685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD17B2 PubMed Links

Gene Details

Gene
HSD17B2
Gene Name
hydroxysteroid 17-beta dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002153.2 190 Silent Mutation TCA,TCG S6S NP_002144.1

View Full Product Details