Product Details
- SNP ID
-
rs11180441
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:75279599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACCAGAGGTTCTCTAATGGACTT[A/G]GACAATAATGCATTGCAAAGGGTCT
- Phenotype
-
MIM: 607724
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPS2
PubMed Links
Gene Details
- Gene
- CAPS2
- Gene Name
- calcyphosine 2
View Full Product Details