Product Details

SNP ID
rs11057348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123767236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTTTTAAATTGTGGTAACAT[A/G]CACACGGCCTGCAATTTACATCTTA
Phenotype
MIM: 611716 MIM: 605884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A2 PubMed Links

Gene Details

Gene
ATP6V0A2
Gene Name
ATPase H+ transporting V0 subunit a2
There are no transcripts associated with this gene.

Gene
DNAH10
Gene Name
dynein axonemal heavy chain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207437.3 Intron NP_997320.2
XM_005253555.3 Intron XP_005253612.1
XM_011538014.2 Intron XP_011536316.1
XM_011538015.2 Intron XP_011536317.1
XM_011538016.2 Intron XP_011536318.1
XM_011538017.2 Intron XP_011536319.1
XM_011538019.2 Intron XP_011536321.1
XM_011538020.2 Intron XP_011536322.1
XM_017018960.1 Intron XP_016874449.1
XM_017018961.1 Intron XP_016874450.1
XM_017018962.1 Intron XP_016874451.1

View Full Product Details