Product Details

SNP ID
rs12126361
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:77284445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTGACTGATTATTGTATTGAC[A/G]TATTGTCAGACAAGTAAATATATCT
Phenotype
MIM: 608009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK5 PubMed Links

Gene Details

Gene
AK5
Gene Name
adenylate kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012093.3 Intron NP_036225.2
NM_174858.2 Intron NP_777283.1
XM_005270739.4 Intron XP_005270796.1
XM_006710572.3 Intron XP_006710635.1
XM_017001008.1 Intron XP_016856497.1
XM_017001009.1 Intron XP_016856498.1
XM_017001010.1 Intron XP_016856499.1
XM_017001011.1 Intron XP_016856500.1
XM_017001012.1 Intron XP_016856501.1

View Full Product Details