Product Details

SNP ID
rs12740132
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:32747030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCAGTTCACTGTGGCCTCTCCA[A/G]GTTCCCTCTGGCCACTCAGGGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1522 PubMed Links

Gene Details

Gene
KIAA1522
Gene Name
KIAA1522
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198972.1 Intron NP_001185901.1
NM_001198973.1 Intron NP_001185902.1
NM_020888.2 Intron NP_065939.2
XM_006710791.3 Intron XP_006710854.1
XM_017001916.1 Intron XP_016857405.1
XM_017001917.1 Intron XP_016857406.1

View Full Product Details