Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007466.2 | Intron | NP_001007467.1 | |||
NM_020245.4 | Intron | NP_064630.2 | |||
XM_011535946.1 | Intron | XP_011534248.1 | |||
XM_017011063.1 | Intron | XP_016866552.1 | |||
XM_017011064.1 | Intron | XP_016866553.1 | |||
XM_017011065.1 | Intron | XP_016866554.1 | |||
XM_017011066.1 | Intron | XP_016866555.1 | |||
XM_017011067.1 | Intron | XP_016866556.1 | |||
XM_017011068.1 | Intron | XP_016866557.1 | |||
XM_017011069.1 | Intron | XP_016866558.1 | |||
XM_017011070.1 | Intron | XP_016866559.1 | |||
XM_017011071.1 | Intron | XP_016866560.1 |