Product Details
- SNP ID
-
rs13407605
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74518265 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTAAGGAAAGAGTCCTTCCCTATG[C/T]AGACTGTGGTTTACCCCATTCTTTC
- Phenotype
-
MIM: 602434
MIM: 610231
MIM: 604240
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AUP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140478168] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AUP1
- Gene Name
- ancient ubiquitous protein 1
There are no transcripts associated with this gene.
- Gene
- DQX1
- Gene Name
- DEAQ-box RNA dependent ATPase 1
- Gene
- PCGF1
- Gene Name
- polycomb group ring finger 1
There are no transcripts associated with this gene.
- Gene
- TLX2
- Gene Name
- T-cell leukemia homeobox 2
There are no transcripts associated with this gene.
View Full Product Details