Product Details

SNP ID
rs12105316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74498203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCTCGGACGGCGCCAGGTACTT[C/T]TGGAAGACGAAGCGCCGCTCCAGCT
Phenotype
MIM: 607164 MIM: 610231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LBX2 PubMed Links

Gene Details

Gene
LBX2
Gene Name
ladybird homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009812.1 766 Silent Mutation CAA,CAG Q103Q NP_001009812.1
NM_001282430.1 766 Silent Mutation CAA,CAG Q107Q NP_001269359.1
Gene
LBX2-AS1
Gene Name
LBX2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PCGF1
Gene Name
polycomb group ring finger 1
There are no transcripts associated with this gene.

Gene
TTC31
Gene Name
tetratricopeptide repeat domain 31
There are no transcripts associated with this gene.

View Full Product Details