Product Details

SNP ID
rs6730473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:71114630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGACATTTGTAAATCAATCTG[A/G]GAAATTTAATTTATGAGCTGTGTAT
Phenotype
MIM: 608419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCEE PubMed Links

Gene Details

Gene
MCEE
Gene Name
methylmalonyl-CoA epimerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032601.3 Intron NP_115990.3
XM_005264613.2 Intron XP_005264670.1

View Full Product Details