Product Details

SNP ID
rs10876955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56924542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGACTGGTGATGAGAAGAACTGT[A/G]CACATTCTCCTTATAACTGTGCCGC
Phenotype
MIM: 609769
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDR9C7 PubMed Links

Gene Details

Gene
SDR9C7
Gene Name
short chain dehydrogenase/reductase family 9C, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148897.2 Intron NP_683695.1

View Full Product Details