Product Details

SNP ID
rs11569712
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:69844175 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
ATTTTTCTCACCCTTTCTCATGAAG[G/T]GCGACAATTTCTGGTTCATAATTTC
Phenotype
MIM: 600043
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SULT1E1 PubMed Links

Gene Details

Gene
SULT1E1
Gene Name
sulfotransferase family 1E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005420.2 871 Missense Mutation CAC,CCC H253P NP_005411.1
XM_011532210.2 871 Intron XP_011530512.1

View Full Product Details