Product Details

SNP ID
hCV31217544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169830441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTTATGTGATTTAACTTATTT[A/T]TTCTTTGACTCTGAGATTATTACTT
Phenotype
MIM: 613831
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
METTL5 PubMed Links

Gene Details

Gene
METTL5
Gene Name
methyltransferase like 5
There are no transcripts associated with this gene.

Gene
UBR3
Gene Name
ubiquitin protein ligase E3 component n-recognin 3 (putative)
There are no transcripts associated with this gene.

View Full Product Details