Product Details

SNP ID
rs12059957
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:67004868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATTTCGAAAAATAGGCATTAAT[A/T]GGAAACATCCCAAATTAGTTAAATA
Phenotype
MIM: 610804
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC35D1 PubMed Links

Gene Details

Gene
SLC35D1
Gene Name
solute carrier family 35 member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015139.2 Intron NP_055954.1
XM_006710478.1 Intron XP_006710541.1
XM_011541070.1 Intron XP_011539372.1

View Full Product Details