Product Details

SNP ID
rs11208197
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:63374043 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGTATTTTCTTTAGTATTTACCA[C/T]AACCACTGAAGACAGGTAATATCTT
Phenotype
MIM: 604566
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ALG6 PubMed Links

Gene Details

Gene
ALG6
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013339.3 Intron NP_037471.2

View Full Product Details