Product Details

SNP ID
rs12408995
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:183637140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAAATGGAGCTAATAATGTGC[A/C]CTTTACAAGATTGTTCTTAGGATTA
Phenotype
MIM: 609908 MIM: 604227 MIM: 605667
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOBEC4 PubMed Links

Gene Details

Gene
APOBEC4
Gene Name
apolipoprotein B mRNA editing enzyme catalytic polypeptide like 4
There are no transcripts associated with this gene.

Gene
ARPC5
Gene Name
actin related protein 2/3 complex subunit 5
There are no transcripts associated with this gene.

Gene
RGL1
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297669.1 Intron NP_001284598.1
NM_001297670.1 Intron NP_001284599.1
NM_001297671.1 Intron NP_001284600.1
NM_001297672.1 Intron NP_001284601.1
NM_015149.4 Intron NP_055964.3
XM_011509339.2 Intron XP_011507641.1
XM_011509341.1 Intron XP_011507643.1
XM_011509342.2 Intron XP_011507644.1
XM_017000756.1 Intron XP_016856245.1

View Full Product Details