Product Details

SNP ID
rs7836130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52540376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGTCTAGGATAATTGTAAAATG[A/G]GCAGGGCCTTGAGAAGTCCCTTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM150A PubMed Links

Gene Details

Gene
FAM150A
Gene Name
family with sequence similarity 150 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207413.3 Intron NP_997296.1

View Full Product Details