Product Details

SNP ID
rs10971456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33276224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGTTTTAAGAGTTACTTGAACCT[C/G]TCCTTTAGAGGAGTGTCTGCTCAGT
Phenotype
MIM: 610900
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHMP5 PubMed Links

Gene Details

Gene
CHMP5
Gene Name
charged multivesicular body protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195536.1 Intron NP_001182465.1
NM_016410.5 Intron NP_057494.3

View Full Product Details