Product Details

SNP ID
rs10971362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33043720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCAATATTGAGAGTGAGGCAA[A/G]AGAGAAAAAAAACCTAGCTTCGGAG
Phenotype
MIM: 602837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJA1 PubMed Links

Gene Details

Gene
DNAJA1
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
There are no transcripts associated with this gene.

Gene
SMU1
Gene Name
DNA replication regulator and spliceosomal factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018225.2 5223 UTR 3 NP_060695.2
XM_005251503.4 5223 UTR 3 XP_005251560.1

View Full Product Details