Product Details

SNP ID
rs12241101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17650544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTAATTCTTGACTTCTGTAGTTG[C/T]AATTTTCTACATTCTAAACAACAAA
Phenotype
MIM: 601899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STAM PubMed Links

Gene Details

Gene
STAM
Gene Name
signal transducing adaptor molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324282.1 Intron NP_001311211.1
NM_001324283.1 Intron NP_001311212.1
NM_001324284.1 Intron NP_001311213.1
NM_001324285.1 Intron NP_001311214.1
NM_001324286.1 Intron NP_001311215.1
NM_001324287.1 Intron NP_001311216.1
NM_001324288.1 Intron NP_001311217.1
NM_001324289.1 Intron NP_001311218.1
NM_003473.3 Intron NP_003464.1
XM_011519695.2 Intron XP_011517997.1
XM_017016685.1 Intron XP_016872174.1
Gene
STAM-AS1
Gene Name
STAM antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details