Product Details

SNP ID
rs12545160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41582626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTTTTTAAAAAGCACAAAGAA[A/G]TAGATAAGGTTGATTTAATCATTCC
Phenotype
MIM: 608143
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPAT4 PubMed Links

Gene Details

Gene
GPAT4
Gene Name
glycerol-3-phosphate acyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178819.3 Intron NP_848934.1
XM_011544392.2 Intron XP_011542694.1
XM_011544393.1 Intron XP_011542695.1
XM_011544394.1 Intron XP_011542696.1
XM_017013042.1 Intron XP_016868531.1
XM_017013043.1 Intron XP_016868532.1
Gene
LOC102723729
Gene Name
uncharacterized LOC102723729
There are no transcripts associated with this gene.

View Full Product Details