Product Details

SNP ID
rs3132594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30647483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCGCGGGAGCGGGAGGGAGGCG[T/C]TGCCGGGCCTGTCGCGCAAGGACGT
Phenotype
MIM: 615556 MIM: 603405
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031722.3 445 Intron NP_001026892.1
NM_001190724.2 445 Intron NP_001177653.1
NM_001254952.2 445 Intron NP_001241881.1
NM_001318762.1 445 Intron NP_001305691.1
NM_001318763.1 445 Intron NP_001305692.1
NM_024909.3 445 Intron NP_079185.2
XM_005249415.2 445 Intron XP_005249472.1
XM_011514918.2 445 Intron XP_011513220.1
XM_017011316.1 445 Intron XP_016866805.1
Gene
C6orf136
Gene Name
chromosome 6 open reading frame 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109938.2 445 Intron NP_001103408.1
NM_001161376.1 445 Silent Mutation CGC,CGT R84R NP_001154848.1
NM_145029.3 445 Intron NP_659466.2
XM_011514384.1 445 Intron XP_011512686.1
Gene
DHX16
Gene Name
DEAH-box helicase 16
There are no transcripts associated with this gene.

View Full Product Details