Product Details

SNP ID
rs13378440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94715890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAAAATCCCAGCATATAGGAAA[G/T]GGAATAGTCTGTGTGAGTGAATAGT
Phenotype
MIM: 604974
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927248 PubMed Links

Gene Details

Gene
LOC101927248
Gene Name
uncharacterized LOC101927248
There are no transcripts associated with this gene.

Gene
SOX21
Gene Name
SRY-box 21
There are no transcripts associated with this gene.

Gene
SOX21-AS1
Gene Name
SOX21 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details