Product Details

SNP ID
rs12176017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25964554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATTCTTAGTCCTCTTTTCTTCA[A/G]TACAAAATGAGATAATAGGGGTTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM38 PubMed Links

Gene Details

Gene
TRIM38
Gene Name
tripartite motif containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006355.4 Intron NP_006346.1
XM_005248799.4 Intron XP_005248856.1
XM_005248800.4 Intron XP_005248857.1
XM_017010175.1 Intron XP_016865664.1

View Full Product Details