Product Details

SNP ID
rs11791502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127703743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAGTGGGTACCAGGGGGTGGCA[C/G]CCACTGGTATTCTCAGCTGGTCTCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP157 PubMed Links

Gene Details

Gene
CFAP157
Gene Name
cilia and flagella associated protein 157
There are no transcripts associated with this gene.

Gene
PTRH1
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002913.1 Intron NP_001002913.1
XM_006716955.3 Intron XP_006717018.1
XM_017014276.1 Intron XP_016869765.1
XM_017014277.1 Intron XP_016869766.1
XM_017014278.1 Intron XP_016869767.1

View Full Product Details