Product Details

SNP ID
rs6478846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128692539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATATTGAGCAATTGCCTTTAGT[C/T]AATAATGGGTTTATAAAACCAAAGA
Phenotype
MIM: 610714 MIM: 600960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
There are no transcripts associated with this gene.

Gene
SET
Gene Name
SET nuclear proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122821.1 Intron NP_001116293.1
NM_001248000.1 Intron NP_001234929.1
NM_001248001.1 Intron NP_001234930.1
NM_003011.3 Intron NP_003002.2
XM_017015013.1 Intron XP_016870502.1
XM_017015014.1 Intron XP_016870503.1
XM_017015015.1 Intron XP_016870504.1
XM_017015016.1 Intron XP_016870505.1

View Full Product Details