Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001035.2 | Intron | NP_001026.2 | |||
XM_006711802.3 | Intron | XP_006711865.1 | |||
XM_006711803.3 | Intron | XP_006711866.1 | |||
XM_006711804.3 | Intron | XP_006711867.1 | |||
XM_006711805.3 | Intron | XP_006711868.1 | |||
XM_006711806.3 | Intron | XP_006711869.1 | |||
XM_006711807.3 | Intron | XP_006711870.1 | |||
XM_006711808.3 | Intron | XP_006711871.1 | |||
XM_006711809.3 | Intron | XP_006711872.1 | |||
XM_006711810.3 | Intron | XP_006711873.1 | |||
XM_017002028.1 | Intron | XP_016857517.1 |