Product Details

SNP ID
rs11169079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49584723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCAGAGCCAAGTCCAGAGCTGA[A/G]CCCAGACTCAGCCTGGACACAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM186B PubMed Links

Gene Details

Gene
FAM186B
Gene Name
family with sequence similarity 186 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032130.2 Intron NP_115506.1
XM_006719625.2 Intron XP_006719688.1
XM_006719626.2 Intron XP_006719689.1
XM_006719627.3 Intron XP_006719690.1
XM_011538796.2 Intron XP_011537098.1
XM_011538797.2 Intron XP_011537099.1
XM_017020008.1 Intron XP_016875497.1

View Full Product Details