Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278682.1 | Intron | NP_001265611.1 | |||
NM_001278684.1 | Intron | NP_001265613.1 | |||
NM_001278686.1 | Intron | NP_001265615.1 | |||
NM_003244.3 | Intron | NP_003235.1 | |||
NM_170695.3 | Intron | NP_733796.2 | |||
NM_173207.2 | Intron | NP_775299.1 | |||
NM_173208.2 | Intron | NP_775300.1 | |||
NM_173209.2 | Intron | NP_775301.1 | |||
NM_173210.2 | Intron | NP_775302.1 | |||
NM_173211.1 | Intron | NP_775303.1 | |||
NM_174886.2 | Intron | NP_777480.1 | |||
XM_011525735.2 | Intron | XP_011524037.1 | |||
XM_017025958.1 | Intron | XP_016881447.1 | |||
XM_017025959.1 | Intron | XP_016881448.1 |