Product Details

SNP ID
rs12456403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:3503365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCATTTGCTAAGTGATAGTAAA[A/C]ATTCTTCAGTGTTTCAGGATTGACT
Phenotype
MIM: 605445
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLGAP1 PubMed Links

Gene Details

Gene
DLGAP1
Gene Name
DLG associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003809.2 Intron NP_001003809.1
NM_001242761.1 Intron NP_001229690.1
NM_001242762.1 Intron NP_001229691.1
NM_001242763.1 Intron NP_001229692.1
NM_001242764.1 Intron NP_001229693.1
NM_001242765.1 Intron NP_001229694.1
NM_001242766.1 Intron NP_001229695.1
NM_001308390.1 Intron NP_001295319.1
NM_004746.3 Intron NP_004737.2
XM_005258171.2 Intron XP_005258228.1
XM_005258172.1 Intron XP_005258229.1
XM_005258173.3 Intron XP_005258230.2
XM_005258174.4 Intron XP_005258231.1
XM_006722367.3 Intron XP_006722430.1
XM_011525770.2 Intron XP_011524072.1
XM_011525771.2 Intron XP_011524073.1
XM_017026080.1 Intron XP_016881569.1
XM_017026081.1 Intron XP_016881570.1
XM_017026082.1 Intron XP_016881571.1
XM_017026083.1 Intron XP_016881572.1
XM_017026084.1 Intron XP_016881573.1
XM_017026085.1 Intron XP_016881574.1

View Full Product Details