Product Details

SNP ID
rs12907108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40706055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAATTTTATTTGATATTTTCT[A/C]TTCCCATTGCACACCTTGTTTTACT
Phenotype
MIM: 179617
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164269.1 Intron NP_001157741.1
NM_001164270.1 Intron NP_001157742.1
NM_002875.4 Intron NP_002866.2
NM_133487.3 Intron NP_597994.3
XM_006720626.3 Intron XP_006720689.1
XM_011521857.2 Intron XP_011520159.1
XM_011521858.2 Intron XP_011520160.1
XM_011521859.2 Intron XP_011520161.1
XM_011521860.2 Intron XP_011520162.1
XM_011521861.2 Intron XP_011520163.1
XM_011521862.2 Intron XP_011520164.1

View Full Product Details