Product Details

SNP ID
rs7163613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40827214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGGCTATTTAACACTTGAGA[G/T]GTGGCCCATGTTGGCTGGGTGCGGT
Phenotype
MIM: 613256
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R14D PubMed Links

Gene Details

Gene
PPP1R14D
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130143.1 Intron NP_001123615.1
NM_017726.7 Intron NP_060196.1
XM_017022372.1 Intron XP_016877861.1

View Full Product Details